Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096895.4 | XP_001096895.3 | 532 | 135 | ||
| GenBank mRNA | JU323271.1 | - | 137 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LY86
Genomic Map