Assay Details
| Gene Symbol: RABGGTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261560.2 | NP_001248489.1 | 336 | 161 | ||
| GenBank mRNA | JU335736.1 | - | 363 | 161 | ||
| JU473413.1 | - | 336 | 161 | |||
| JV490939.1 | - | 367 | 161 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RABGGTB
Genomic Map