Assay Details
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111059.2 | XP_014966545.1 | 1014 | 72 | ||
XM_015111060.2 | XP_014966546.1 | 1014 | 72 | |||
XM_015111061.2 | XP_014966547.1 | 746 | 72 | |||
GenBank mRNA | JU330992.1 | - | 634 | 72 | ||
JU477771.1 | - | 644 | 72 | |||
JV665243.1 | - | 645 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
