Assay Details
Gene Symbol: DNAJC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257595.1 | NP_001244524.1 | 454 | 103 | ||
XM_028831426.1 | XP_028687259.1 | 536 | 103 | |||
GenBank mRNA | JU321934.1 | - | 454 | 103 | ||
JU473215.1 | - | 458 | 103 | |||
JV043655.1 | - | 453 | 103 | |||
JV043656.1 | - | 453 | 103 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
