Assay Details
Gene Symbol: IRAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114866.2 | XP_014970352.2 | 1360 | 66 | ||
XM_015114867.2 | XP_014970353.2 | 1813 | 66 | |||
XM_015114868.2 | XP_014970354.2 | 1810 | 66 | |||
XM_015114870.2 | XP_014970356.2 | 1729 | 66 | |||
XM_015114871.2 | XP_014970357.2 | 1726 | 66 | |||
XM_015114873.2 | XP_014970359.2 | 1363 | 66 | |||
XM_015114874.2 | XP_014970360.2 | 1573 | 66 | |||
XM_015114875.2 | XP_014970361.2 | 1807 | 66 | |||
XM_028833687.1 | XP_028689520.1 | 1363 | 66 | |||
XM_028833688.1 | XP_028689521.1 | 1748 | 66 | |||
XM_028833689.1 | XP_028689522.1 | 1388 | 66 | |||
GenBank mRNA | JU337701.1 | - | 1256 | 66 | ||
JV705125.1 | - | 1379 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
