Assay Details
| Gene Symbol: GLRX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090479.4 | XP_001090479.1 | 693 | 89 | ||
| XM_028827019.1 | XP_028682852.1 | 754 | 89 | |||
| GenBank mRNA | JV531063.1 | - | 661 | 89 | ||
| JV603659.1 | - | 674 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GLRX3
Genomic Map