Assay Details
| Gene Symbol: PHACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090338.4 | XP_001090338.1 | 1841 | 96 | ||
| XM_001090453.4 | XP_001090453.1 | 1540 | 96 | |||
| XM_001090568.4 | XP_001090568.1 | 1597 | 96 | |||
| XM_002803874.3 | XP_002803920.1 | 1786 | 96 | |||
| XM_015137221.2 | XP_014992707.1 | 1826 | 96 | |||
| XM_015137223.2 | XP_014992709.1 | 1655 | 96 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PHACTR2
Genomic Map