Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143226.2 | XP_014998712.1 | 1645 | 104 | ||
| XM_015143227.2 | XP_014998713.1 | 1651 | 104 | |||
More Information
Set Membership: |
|

SLC39A2
Genomic Map