Assay Details
| Gene Symbol: FEZF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844307.1 | XP_028700140.1 | 1823 | 95 | ||
| XM_028844308.1 | XP_028700141.1 | 1533 | 95 | |||
| XM_028844309.1 | XP_028700142.1 | 1794 | 95 | |||
More Information
Set Membership: |
|

FEZF2
Genomic Map