Assay Details
| Gene Symbol: TMEM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193979.3 | NP_001180908.1 | 996 | 90 | ||
| XM_015150312.2 | XP_015005798.1 | 1008 | 90 | |||
| XM_015150313.2 | XP_015005799.1 | 1109 | 90 | |||
| XM_015150314.2 | XP_015005800.1 | 1109 | 90 | |||
| GenBank mRNA | JU321372.1 | - | 977 | 90 | ||
| JU470924.1 | - | 965 | 90 | |||
| JV044164.1 | - | 90 | ||||
| JV638773.1 | - | 984 | 90 | |||
| JV704698.1 | - | 990 | 90 | |||
More Information
Set Membership: |
|

TMEM184B
Genomic Map