Assay Details
| Gene Symbol: PHACTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260596.1 | NP_001247525.1 | 1705 | 72 | ||
| XM_015135538.2 | XP_014991024.2 | 2316 | 72 | |||
| XM_015135539.2 | XP_014991025.1 | 2211 | 72 | |||
| XM_015135544.2 | XP_014991030.1 | 2035 | 72 | |||
| XM_015135546.2 | XP_014991032.1 | 1844 | 72 | |||
| XM_015135548.2 | XP_014991034.1 | 2124 | 72 | |||
| XM_015135550.2 | XP_014991036.1 | 1911 | 72 | |||
| XM_028846697.1 | XP_028702530.1 | 1799 | 72 | |||
| XM_028846698.1 | XP_028702531.1 | 1763 | 72 | |||
| XM_028846699.1 | XP_028702532.1 | 1447 | 72 | |||
| GenBank mRNA | JU333408.1 | - | 1705 | 72 | ||
| JV602399.1 | - | 1712 | 72 | |||
More Information
Set Membership: |
|

PHACTR1
Genomic Map