Assay Details
Gene Symbol: PAK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261653.1 | NP_001248582.1 | 2009 | 99 | ||
XM_015141858.2 | XP_014997344.1 | 2732 | 99 | |||
XM_015141860.2 | XP_014997346.1 | 2962 | 99 | |||
XM_015141861.2 | XP_014997347.1 | 2453 | 99 | |||
XM_015141863.1 | XP_014997349.1 | 2366 | 99 | |||
XM_015141864.2 | XP_014997350.1 | 2212 | 99 | |||
XM_028850281.1 | XP_028706114.1 | 2327 | 99 | |||
XM_028850282.1 | XP_028706115.1 | 2411 | 99 | |||
GenBank mRNA | JU335534.1 | - | 2083 | 99 | ||
JU335535.1 | - | 2083 | 99 | |||
JV047702.1 | - | 2009 | 99 | |||
JV047703.1 | - | 2009 | 99 | |||
JV047704.1 | - | 2009 | 99 | |||
JV047705.1 | - | 2009 | 99 | |||
JV601293.1 | - | 2317 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
