Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194811.4 | NP_001181740.2 | 894 | 55 | ||
| XM_028835646.1 | XP_028691479.1 | 893 | 55 | |||
| GenBank mRNA | JU336357.1 | - | 783 | 55 | ||
| JV529351.1 | - | 813 | 55 | |||
| JV593398.1 | - | 847 | 55 | |||
| JV628514.1 | - | 858 | 55 | |||
More Information
Set Membership: |
|

DHRS11
Genomic Map