Assay Details
| Gene Symbol: GABRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260702.1 | NP_001247631.1 | 243 | 87 | ||
| XM_015141717.2 | XP_014997203.1 | 431 | 87 | |||
| XM_015141718.2 | XP_014997204.1 | 536 | 87 | |||
| GenBank mRNA | JU333822.1 | - | 243 | 87 | ||
| JV711345.1 | - | 277 | 87 | |||
More Information
Set Membership: |
|

GABRB3
Genomic Map