Assay Details
| Gene Symbol: MOGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109499.4 | XP_001109499.1 | 784 | 107 | ||
| GenBank mRNA | JU325904.1 | - | 195 | 107 | ||
| JU470475.1 | - | 492 | 107 | |||
| JV044169.1 | - | 488 | 107 | |||
| JV628311.1 | - | 504 | 107 | |||
| JV635420.1 | - | 498 | 107 | |||
More Information
Set Membership: |
|

MOGS
Genomic Map