Assay Details
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257599.2 | NP_001244528.1 | 267 | 67 | ||
XM_015137080.2 | XP_014992566.1 | 400 | 67 | |||
XM_015137081.2 | XP_014992567.1 | 396 | 67 | |||
XM_015137082.2 | XP_014992568.1 | 425 | 67 | |||
XM_015137083.2 | XP_014992569.1 | 399 | 67 | |||
XM_028846725.1 | XP_028702558.1 | 199 | 67 | |||
GenBank mRNA | JU333907.1 | - | 270 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
