Assay Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265927.1 | NP_001252856.1 | 527 | 116 | ||
| XM_015126712.2 | XP_014982198.1 | 618 | 116 | |||
| GenBank mRNA | JU320694.1 | - | 540 | 116 | ||
| JU472808.1 | - | 516 | 116 | |||
| JU472809.1 | - | 516 | 116 | |||
| JV045890.1 | - | 527 | 116 | |||
More Information
Set Membership: |
|

EMC8
Genomic Map