Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193906.2 | NP_001180835.1 | 1221 | 83 | ||
XM_015140836.2 | XP_014996322.1 | 1333 | 83 | |||
XM_015140837.2 | XP_014996323.1 | 1333 | 83 | |||
XM_015140838.2 | XP_014996324.1 | 1312 | 83 | |||
XM_015140839.2 | XP_014996325.1 | 1333 | 83 | |||
XM_028849181.1 | XP_028705014.1 | 1312 | 83 | |||
GenBank mRNA | JU323418.1 | - | 1221 | 83 | ||
JU323420.1 | - | 1221 | 83 | |||
JU478038.1 | - | 1308 | 83 | |||
JU478039.1 | - | 1308 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
