Assay Details
| Gene Symbol: VCPKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266438.1 | NP_001253367.1 | 379 | 67 | ||
| XM_015143634.2 | XP_014999120.2 | 589 | 67 | |||
| XM_015143635.2 | XP_014999121.2 | 589 | 67 | |||
| GenBank mRNA | JU322986.1 | - | 380 | 67 | ||
| JU477203.1 | - | 379 | 67 | |||
More Information
Set Membership: |
|

VCPKMT
Genomic Map