Assay Details
| Gene Symbol: RSBN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257886.1 | NP_001244815.1 | 1627 | 82 | ||
| GenBank mRNA | JU333418.1 | - | 1627 | 82 | ||
| JU482565.1 | - | 1618 | 82 | |||
More Information
Set Membership: |
|

RSBN1L
Genomic Map