Assay Details
| Gene Symbol: FRRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361042.2 | NP_001347971.2 | 632 | 78 | ||
| XM_028844175.1 | XP_028700008.1 | 561 | 78 | |||
| XM_028844177.1 | XP_028700010.1 | 622 | 78 | |||
| GenBank mRNA | JV651171.1 | - | 227 | 78 | ||
More Information
Set Membership: |
|

FRRS1
Genomic Map