Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102507.4 | XP_001102507.2 | 628 | 140 | ||
| XM_015124105.2 | XP_014979591.2 | 853 | 140 | |||
| GenBank mRNA | JU325273.1 | - | 374 | 140 | ||
| JU476190.1 | - | 623 | 140 | |||
| JV717908.1 | - | 613 | 140 | |||
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102228.4 | XP_001102228.1 | 591 | 140 | ||
| XM_002801251.3 | XP_002801297.1 | 654 | 140 | |||
| XM_028839959.1 | XP_028695792.1 | 671 | 140 | |||
| XM_028839960.1 | XP_028695793.1 | 856 | 140 | |||
| GenBank mRNA | JV654970.1 | - | 606 | 140 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DMAC2
Genomic Map