Assay Details
| Gene Symbol: LAMTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194634.3 | NP_001181563.1 | 424 | 99 | ||
| XM_028832842.1 | XP_028688675.1 | 405 | 99 | |||
| GenBank mRNA | JU319621.1 | - | 405 | 99 | ||
| JU476172.1 | - | 343 | 99 | |||
| JV045650.1 | - | 310 | 99 | |||
| JV601383.1 | - | 410 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LAMTOR1
Genomic Map