Assay Details
| Gene Symbol: MAP3K11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257468.1 | NP_001244397.1 | 2703 | 88 | ||
| GenBank mRNA | JU321005.1 | - | 2703 | 88 | ||
| JU473336.1 | - | 2701 | 88 | |||
| JV044425.1 | - | 2615 | 88 | |||
More Information
Set Membership: |
|

MAP3K11
Genomic Map