Assay Details
| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114945.1 | NP_001108417.1 | 239 | 72 | ||
| XM_015135848.2 | XP_014991334.1 | 409 | 72 | |||
| XM_015135849.2 | XP_014991335.1 | 415 | 72 | |||
| GenBank mRNA | JU322719.1 | - | 508 | 72 | ||
| JU472298.1 | - | 501 | 72 | |||
| JV045782.1 | - | 396 | 72 | |||
More Information
Set Membership: |
|

POLR1H
Genomic Map