Assay Details
| Gene Symbol: TGFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266518.1 | NP_001253447.1 | 858 | 104 | ||
| XM_015114984.2 | XP_014970470.1 | 3040 | 104 | |||
| GenBank mRNA | JU320953.1 | - | 858 | 104 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TGFB2
Genomic Map