Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152926.2 | XP_015008412.1 | 586 | 60 | ||
| XM_015152927.2 | XP_015008413.1 | 720 | 60 | |||
| GenBank mRNA | JU319970.1 | - | 615 | 60 | ||
| JU471167.1 | - | 633 | 60 | |||
| JV047642.1 | - | 595 | 60 | |||
More Information
Set Membership: |
|

WSB2
Genomic Map