Assay Details
Gene Symbol: TCP11L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194784.2 | NP_001181713.1 | 1030 | 96 | ||
XM_015114501.2 | XP_014969987.1 | 1083 | 96 | |||
XM_015114502.2 | XP_014969988.1 | 743 | 96 | |||
GenBank mRNA | JU321631.1 | - | 725 | 96 | ||
JU476280.1 | - | 699 | 96 | |||
JV529837.1 | - | 754 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
