Assay Details
| Gene Symbol: MRPS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261058.1 | NP_001247987.1 | 533 | 81 | ||
| GenBank mRNA | JU334774.1 | - | 564 | 81 | ||
| JU475816.1 | - | 533 | 81 | |||
| JV583507.1 | - | 597 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPS7
Genomic Map