Assay Details
| Gene Symbol: RGS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260517.1 | NP_001247446.1 | 372 | 90 | ||
| XM_028853363.1 | XP_028709196.1 | 184 | 90 | |||
| GenBank mRNA | JU336789.1 | - | 472 | 90 | ||
| JV048487.1 | - | 372 | 90 | |||
| JV703642.1 | - | 482 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028853363.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGS5
Genomic Map