Assay Details
| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135848.2 | XP_014991334.1 | 160 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015135848.2 transcripts.
Set Membership: |
|

POLR1H
Genomic Map