Assay Details
Gene Symbol: TMBIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266529.1 | NP_001253458.1 | 540 | 68 | ||
XM_015110970.2 | XP_014966456.1 | 929 | 68 | |||
XM_015110971.2 | XP_014966457.1 | 765 | 68 | |||
XM_028830423.1 | XP_028686256.1 | 560 | 68 | |||
GenBank mRNA | JU335879.1 | - | 552 | 68 | ||
JU476761.1 | - | 550 | 68 | |||
JV047908.1 | - | 540 | 68 | |||
JV658586.1 | - | 547 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
