Assay Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133301.2 | XP_014988787.1 | 445 | 73 | ||
| GenBank mRNA | JU324186.1 | - | 410 | 73 | ||
| JV048402.1 | - | 388 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM120A
Genomic Map