Assay Details
| Gene Symbol: GNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266130.1 | NP_001253059.1 | 136 | 56 | ||
| GenBank mRNA | JU320677.1 | - | 136 | 56 | ||
| JV527828.1 | - | 461 | 56 | |||
More Information
Set Membership: |
|

GNB3
Genomic Map