Assay Details
| Gene Symbol: BHMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140323.2 | XP_014995809.1 | 293 | 72 | ||
| GenBank mRNA | JV590323.1 | - | 194 | 72 | ||
| JV704367.1 | - | 271 | 72 | |||
More Information
Set Membership: |
|

BHMT
Genomic Map