Assay Details
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261314.1 | NP_001248243.1 | 458 | 133 | ||
XM_015134720.2 | XP_014990206.1 | 557 | 133 | |||
XM_015134721.2 | XP_014990207.1 | 577 | 133 | |||
GenBank mRNA | JU320227.1 | - | 458 | 133 | ||
JV490668.1 | - | 484 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
