Assay Details
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114958.1 | NP_001108430.1 | 643 | 96 | ||
GenBank mRNA | JU323170.1 | - | 662 | 96 | ||
JU473200.1 | - | 664 | 96 | |||
JV071116.1 | - | 603 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
