Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131586.2 | XP_014987072.2 | 1808 | 119 | ||
| XM_015131588.2 | XP_014987074.2 | 1809 | 119 | |||
| XM_028843862.1 | XP_028699695.1 | 1337 | 119 | |||
| XM_028843863.1 | XP_028699696.1 | 1809 | 119 | |||
| GenBank mRNA | JU341356.1 | - | 502 | 119 | ||
More Information
Set Membership: |
|

NAALADL2
Genomic Map