Assay Details
| Gene Symbol: PLD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118731.2 | XP_014974217.1 | 550 | 93 | ||
| GenBank mRNA | JV054199.1 | - | 76 | 93 | ||
| JV636016.1 | - | 434 | 93 | |||
More Information
Set Membership: |
|

PLD6
Genomic Map