Assay Details
Gene Symbol: HMBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266660.1 | NP_001253589.1 | 972 | 121 | ||
XM_015115888.2 | XP_014971374.1 | 1098 | 121 | |||
XM_015115889.2 | XP_014971375.1 | 3472 | 121 | |||
XM_028832601.1 | XP_028688434.1 | 954 | 121 | |||
GenBank mRNA | JU334262.1 | - | 981 | 121 | ||
JU475372.1 | - | 972 | 121 | |||
JV045500.1 | - | 945 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
