Assay Details
| Gene Symbol: HEPACAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090392.4 | XP_001090392.2 | 1028 | 106 | ||
| XM_015134263.2 | XP_014989749.1 | 1034 | 106 | |||
| XM_015134264.1 | XP_014989750.1 | 697 | 106 | |||
| XM_028846059.1 | XP_028701892.1 | 1465 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HEPACAM2
Genomic Map