Assay Details
| Gene Symbol: IL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042756.1 | NP_001036221.1 | 467 | 95 | ||
| GenBank mRNA | U19845.1 | - | 467 | 95 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IL1B
Genomic Map