Assay Details
| Gene Symbol: FASLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032838.1 | NP_001028010.1 | 389 | 118 | ||
| GenBank mRNA | AB035139.1 | - | 347 | 118 | ||
| AF344856.1 | - | 389 | 118 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FASLG
Genomic Map