Assay Details
| Gene Symbol: CCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032850.1 | NP_001028022.1 | 187 | 74 | ||
| XM_015119022.2 | XP_014974508.1 | 275 | 74 | |||
| GenBank mRNA | AF449268.1 | - | 187 | 74 | ||
| AF457194.1 | - | 95 | 74 | |||
| DQ913691.1 | - | 275 | 74 | |||
| DQ913693.1 | - | 274 | 74 | |||
| DQ913697.1 | - | 275 | 74 | |||
| DQ913698.1 | - | 274 | 74 | |||
| DQ913699.1 | - | 275 | 74 | |||
| DQ913700.1 | - | 274 | 74 | |||
| DQ913701.1 | - | 275 | 74 | |||
| DQ913702.1 | - | 275 | 74 | |||
| DQ913703.1 | - | 275 | 74 | |||
| DQ913709.1 | - | 275 | 74 | |||
| DQ913711.1 | - | 275 | 74 | |||
| DQ913715.1 | - | 275 | 74 | |||
| DQ913730.1 | - | 274 | 74 | |||
| JU472169.1 | - | 231 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCL5
Genomic Map