Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193822.1 | NP_001180751.1 | 628 | 90 | ||
GenBank mRNA | JV581004.1 | - | 625 | 90 | ||
JV714287.1 | - | 603 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
