Assay Details
| Gene Symbol: BBC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124339.2 | XP_014979825.1 | 771 | 105 | ||
| XM_015124340.2 | XP_014979826.1 | 2463 | 105 | |||
| GenBank mRNA | JV613260.1 | - | 218 | 105 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BBC3
Genomic Map