Assay Details
Gene Symbol: AKIRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136918.2 | XP_014992404.1 | 1021 | 108 | ||
GenBank mRNA | JU319914.1 | - | 954 | 108 | ||
JU471356.1 | - | 972 | 108 | |||
JV045336.1 | - | 741 | 108 | |||
JV567767.1 | - | 966 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
