Assay Details
| Gene Symbol: CCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032858.1 | NP_001028030.1 | 860 | 66 | ||
| XM_015131075.2 | XP_014986561.1 | 1019 | 66 | |||
| GenBank mRNA | AF017282.1 | - | 860 | 66 | ||
| JV703763.1 | - | 931 | 66 | |||
More Information
Set Membership: |
|

CCR1
Genomic Map