Assay Details
| Gene Symbol: DEFB118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037344.1 | NP_001032421.1 | 81 | 79 | ||
| GenBank mRNA | AF207834.1 | - | 81 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEFB118| Gene Symbol: DEFB118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037344.1 | NP_001032421.1 | 81 | 79 | ||
| GenBank mRNA | AF207834.1 | - | 81 | 79 | ||
Set Membership: |
|