Assay Details
| Gene Symbol: NPY2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032832.1 | NP_001028004.1 | 1087 | 110 | ||
| XM_028847921.1 | XP_028703754.1 | 2134 | 110 | |||
| XM_028847922.1 | XP_028703755.1 | 1597 | 110 | |||
| GenBank mRNA | AF303090.1 | - | 1087 | 110 | ||
More Information
Set Membership: |
|

NPY2R
Genomic Map